missing translation for 'onlineSavingsMsg'
Learn More

Clobetasol propionate, ≥98%, MilliporeSigma™ Supelco™

Clobetasol propionate is a novel and highly potent, topical corticosteroid, which is being assessed for its utility in the treatment of bullous pemphigoid.

$117.69 - $772.34

Specifications

Packaging Glass Bottle
CAS 25122-46-7
Physical Form Neat
Linear Formula C25H32 ClFO5
MDL Number MFCD00058499
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012185
View Documents
Merck Emd Millipore
C8037100MG
100 mg N/A
Only null left
 
111012186
View Documents
Merck Emd Millipore
C80371G
1 g N/A
Only null left
 
Description

Description

Specifications

Specifications

Glass Bottle
Neat
MFCD00058499
466.97
Analytical Standard
25122-46-7
C25H32 ClFO5
Analytical Standard
≥98%
Videos
SDS
Documents

Documents