missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clobetasol propionate

Chemical Identifiers

CAS 25122-46-7
Chemical Name or Material Cloβsol propionate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466820010
View Documents
Thermo Scientific™
466820010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466820050
View Documents
Thermo Scientific™
466820050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

25122-46-7
Cloβsol propionate

Specifications

25122-46-7
Solubility aq. soln.: insoluble.
Cloβsol propionate
1 g
191.0°C to 195.0°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement May damage fertility or the unborn child.

May cause damage to organs through prolonged or repeated exposure.

May cause long lasting harmful effects to aquatic life.

GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 246-634-3

SDS
Documents

Documents

RUO – Research Use Only