missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clobetasol propionate

Supplier:  Thermo Scientific™ 466820050

 View more versions of this product

Catalog No. AC466820050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cloβsol propionate
5 g
191.0°C to 195.0°C
25122-46-7
Solubility aq. soln.: insoluble.

Chemical Identifiers

25122-46-7
Cloβsol propionate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement May damage fertility or the unborn child.

May cause damage to organs through prolonged or repeated exposure.

May cause long lasting harmful effects to aquatic life.

GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 246-634-3

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only