missing translation for 'onlineSavingsMsg'
Learn More

Medisca Clobetasol 17-Propionate USP (Micronized)

Catalog No. 14185411
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185411 5 g
14185134 100 g
14185194 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14185411 Supplier Medisca Supplier No. 037203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clobetasol 17-Propionate USP is a white to cream, crystalline powder. Micronized with a particle size distribution of 99% ≤ 50 μm.

Clobetasol 17-Propionate USP is a white to cream, crystalline powder. Micronized with a particle size distribution of 99% ≤ 50 μm.

Alternative name(s): clobetasol propionate, Clobetasone propionate

Specifications

Chemical Name or Material Clobetasol 17-Propionate USP (Micronized)
Packaging Bottle, 30mL, Amber Glass, Wide Mouth Packer, 28-400
CAS 25122-46-7
Type Drug Standard
Molecular Weight (g/mol) 4669738
Quantity 5 g
Percent Purity 97 to 102 %
Grade USP