Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
$78.07 - $130.75
| Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
|
14-488-146
|
Thermo Scientific™
94410219 |
ClipTip Empty Rack, Small |
Pack of 10 Each for $78.07
|
|
|||||
|
04100505
|
Thermo Scientific™
94410619 |
ClipTip Empty Rack |
Pack of 8 Each for $130.75
|
|
|||||
|
14-488-011
|
Thermo Scientific™
94410519 |
ClipTip Empty Rack, Medium |
Pack of 10 Each for $104.01
|
|
|||||
|
14-488-012
|
Thermo Scientific™
94410819 |
ClipTip Empty Rack, Large |
Pack of 8 Each for $104.01
|
|
|||||
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
- Small Rack: For use with ClipTip 20 and 50 Pipette Tips
- Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
- Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
| ClipTip Empty Rack | |
| ClipTip |