Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Supplier: Thermo Scientific™ 94410619
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
- Small Rack: For use with ClipTip 20 and 50 Pipette Tips
- Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
- Large Rack: For use with ClipTip 1000 and 1250 Pipette Tips
- CT300 Ext Rack: For use with ClipTip 300 Ext Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
| ClipTip Empty Rack | |
| ClipTip Empty Rack | |
| ClipTip 300 Ext Pipette Tip | |
| ClipTip |