missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. p-7125366
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C22565G 5 g
C225625G 25 g
2 options

Catalog No. C22565G

Supplier: TCI America C22565G

Only null left

Chemical Identifiers

CAS 58207-19-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD06411091
InChI Key AUODDLQVRAJAJM-NFOLQJLNSA-N
Synonym clindamycin hydrochloride monohydrate
PubChem CID 131632848
IUPAC Name (2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 5 g
Formula Weight 461.44
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Clindamycin Hydrochloride Monohydrate
EINECSNumber (8)-0464
RTECSNumber GF2275000
TSCA No