missing translation for 'onlineSavingsMsg'
Learn More

Clidinium bromide, MilliporeSigma™ Supelco™

Catalog No. 111013996
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013996 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013996 Supplier Merck Emd Millipore Supplier No. C04141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clidinium bromide is an anticholinergic, antisecretory and antispasmodic drug, widely used in treating anxiety-related conditions and spastic colon by inhibiting the action of parasympathetic innervations thus decreasing the secretions of stomach acid and slowing the intestines.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 3485-62-9
Physical Form Neat
Linear Formula C22H26 BrNO3
MDL Number MFCD00078996
Synonym 3-Hydroxy-1-methylquinuclidinium bromide benzilate; 3-[(Hydroxydiphenylacetyl)oxy]-1-methyl-1-azoniabicyclo[2.2.2 ]octane bromide
Type Analytical Standard
Quantity 1 g
Formula Weight 432.35
Grade Analytical Standard