missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ sDIBO Alkynes for copper-free click chemistry
GREENER_CHOICE

Catalog No. c10407
Encompass
Change view
Click to view available options
Label or Dye:
Alexa Fluor™ 488
Alexa Fluor™ 594
Alexa Fluor™ 647
Reactive Moiety:
DIBO Alkyne
sDIBO Alkyne
Molecular Weight (g/mol):
∼1200
∼1400
∼1900
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Reactive Moiety Molecular Weight (g/mol)
C10407 Alexa Fluor™ 594 DIBO Alkyne ∼1200
C20020 Alexa Fluor™ 488 sDIBO Alkyne ∼1400
C20022 Alexa Fluor™ 647 sDIBO Alkyne ∼1900
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C10407 Supplier Invitrogen™ Supplier No. C10407

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience copper-free 'click chemistry' with Click-IT™ Alexa Fluor™, and sDIBO Alkyne reagents, which are ideal for surface labeling of live cells and minimize damage to enzymes and fluorescent proteins.

Achieve efficient labeling of azides with Click-It sDIBO Alkynes. Our Click-IT sDIBO alkynes enable the labeling of azido labeled targets without the need for copper catalysis and are available with a variety of Alexa Fluor or biotin labels, as well as amine or thiol reactive groups for creating your own conjugates. Macromolecules that have been azido-modified enzymatically, chemically, or metabolically can now be labeled easily, yielding soluble bioconjugates.

Click-iT sDIBO Alkyne labels react with azides via a copper-free Click chemistry reaction to form a stable triazole linkage with an Alexa Fluor, Biotin, or label of your choice. Copper-free click reactions using our sDIBO alkynes are ideal for surface labeling of live cells; they also minimize damage to fluorescent proteins like GFP or R-PE. For intracellular labeling, the nonspecific binding of DIBO alkynes creates a background signal, and a labeling strategy using picolyl azides is the preferred solution in these instances.

Click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background. Macromolecules that have been azide-modified enzymatically, chemically, or metabolically can be labeled more easily, yielding more soluble bioconjugates with improved biological labeling utility.

To make a concentrated stock solution from 2–5 mM, dissolve the dye-alkyne in high quality anhydrous DMSO. Store frozen, desiccated, and protected from light. Click-iT SDP Ester sDIBO alkyne is susceptible to hydrolysis, so prepare the stock solution fresh on the day of the experiment.

Specifications

Quantity 500 μg
Kit Contents Contains 500 μg of Alexa Fluor 594 DIBO Alkyne
Molecular Weight (g/mol) ∼1200
Filter Type Texas Red
Content And Storage Store at ≤-20°C. Desiccation recommended. Protect material from long-term exposure to light; may be exposed to light for short periods of time
Flow Cytometer Laser Lines Yellow
Labeling Method Click Chemistry
Shipping Condition Dry Ice
Reactive Moiety DIBO Alkyne
Label or Dye Alexa Fluor™ 594
Color Red-Orange
Product Line Click-iT
Emission 590 nm
Excitation 617 nm
Labeling Target Azide
Show More Show Less
What concentration and solvent do you recommend for making a stock solution of Click-iT sDIBO Alkynes for copper-free click chemistry?

For Click-iT sDIBO Alkynes for coper-free click chemistry (Cat. Nos. C10407, C20020, C20021, C20022, C20023, C20025), we recommend a 2-5 mM stock solution in DMSO.