missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ Plus Alexa Fluor™ 555 Picolyl Azide Toolkit

Catalog No. C10642
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10642 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10642 Supplier Invitrogen™ Supplier No. C10642
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Click-iT Plus Alexa Fluor 555 Picolyl Azide Toolkit

Click-iT™ Plus Alexa Fluor™ picolyl azide toolkits contain everything you need to perform copper-catalyzed click reactions with compounds that are sensitive to copper, while retaining all of the benefits of the standard azide/alkyne click reaction. Picolyl azides react efficiently with chelated copper, so free copper in the click reaction can be minimized, protecting against undesired copper side reactions with proteins (e.g., GFP, RPE), nucleic acids (e.g., RNA, oligos), and even small molecules (e.g., phalloidin).

  • Flexible—optimize your own Click–iT™ reaction for the detection or labeling of any alkyne-containing biomolecule in vitro or in cells and tissue samples
  • Bright and sensitive—superior fluorescent Alexa Fluor™ dyes combined with the efficiency of picolyl azide
  • Convenient—contains all the buffers and reagents needed to create the optimal click reaction

    Each Click-iT™ Plus Picolyl Azide Toolkit includes 100μg of Alexa Fluor™ dye-picolyl azide and all of the reagents required to create a reaction cocktail with the optimal copper and chelate concentration for any click chemistry-based labeling or detection application. Guidelines for designing and optimizing the picolyl azide/alkyne click reaction of your choice are provided in the product manual. Additionally Click-iT™ Plus picolyl azides offer increased sensitivity and faster reaction times compared to standard azides using identical reaction conditions.
  • Specifications

    Product Type 555 Picolyl Azide Toolkit
    Quantity 1 kit
    Content And Storage
    • 1 x 100 μg Alexa Fluor™ dye-picolyl azide
    • 1 x 4 mL Click-iT™ reaction buffer
    • 1 x 1.5 mL copper (II) sulfate
    • 1 x 500 μL copper protectant
    • 1 Click-iT™ buffer additive
    Each k
    Product Line Click-iT
    Label or Dye Alexa Fluor 555
    Label Type Alexa Fluor Dyes
    What are the main characteristics of a Click-iT reaction?

    Click reactions have several general characteristics: the reaction is efficient, no extreme temperatures or solvents are required, the reaction is complete within 30 minutes, the components of the reaction are bio-inert, and perhaps most importantly, no side reactions occur-the label and detection tags react selectively and specifically with one another. This final point is a key advantage of this powerful detection technique; it is possible to apply click chemistry-labeled molecules to complex biological samples and detect them with unprecedented sensitivity due to the extremely low background of the reaction.

    I will be performing a cell proliferation assay using Click-iT EdU kit. At what point can I stop overnight, or do I have to perform all the steps continuously?

    One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.

    Can I combine Click-iT or Click-iT Plus reactions with phalloidin conjugates used for actin staining?

    We do not recommend using phalloidin conjugates for staining actin in combination with traditional Click-iT or Click-iT Plus reactions since phalloidin is extremely sensitive to the presence of copper.

    For staining actin in combination with traditional Click-iT or Click-iT Plus reactions, we recommend using anti-α-actin antibodies for staining actin in the cytoskeleton. You can find a list of our actin antibodies here.

    Another option would be to use the Click-iT Plus Alexa Fluor Picolyl Azide Toolkit (Cat. Nos. C10641, C10642, C10643). These Click-iT Plus toolkits provide Copper and Copper protectant separately which makes it easier to titrate the copper concentration to obtain optimal labeling with minimal copper-mediated damage. You may need to optimize the click reaction with the lowest possible concentration of copper and then perform the phalloidin staining.

    For Research Use Only. Not for use in diagnostic procedures.