missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit
GREENER_CHOICE

Catalog No. C10428
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10428 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10428 Supplier Invitrogen™ Supplier No. C10428
  • ONLINE EXCLUSIVE PRICE
    $1,158.00
    $949.56 / Each

    Save $208.44

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit

The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit provides a fast, sensitive, non-toxic, and non-radioactive method for the detection of nascent protein synthesis utilizing fluorescence microscopy, high-content imaging, or flow cytometry. Included in the kit are L-homopropargylglycine (HPG), an amino acid analog of methionine containing an alkyne moiety, and Alexa Fluor™ 488 azide. The HPG is fed to cultured cells and incorporated into proteins during active protein synthesis. Addition of the Alexa Fluor™ 488 azide leads to a chemoselective ligation or 'click' reaction between the green fluorescent azide and the alkyne, allowing the modified proteins to be detected by imaged-based analysis.

  • Non-radioactive alternative—an alternative to the traditional 35S-methionine
  • Visualize bulk protein dynamics—fluorescent tagging of proteins allows their localization to be determined, including aggregation
  • Specificity—selective, specific reaction between label and detection tags
  • Stability —product contains an irreversible, covalent bond
  • Multiplex-enabled—use in conjuction with Click™-iT AHA (azide amino acid and alkyne dye) to detect spatial and temporal differences
  • Applicability to biological samples—easy detection; high sensitivity and low background, regardless of complexity

    The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit has been successfully tested in HeLa, A549, and U-2 OS cells with a variety of reagents that inhibit protein synthesis, including cycloheximide and anisomycin. The applicability of these probes to monitor protein degradation has also been shown using inhibitors of the proteasome (MG132 and Bortezomib) and blockers of autophagy (chloroquine) in HeLa cells.

    Additionally, due to differences in Click-iT™ chemistry between the Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit and Click™-iT AHA with Alexa Fluor™ 594 Alkyne, these reagents can be used in conjunction for spatial or temporal determination of differences in nascent protein synthesis.
  • Specifications

    Color Green
    Content And Storage
    • Store at 2°C to 6°C
    • DO NOT FREEZE
    • Protect material from long-term exposure to light, but may be exposed to light for short periods of time.
    Detection Method Fluorescence
    For Use With (Application) Protein Synthesis Assay
    For Use With (Equipment) Fluorescence Microscope, Fluorescent Imager
    Product Type Protein Synthesis Assay Kit
    Dye Type Proteins (Nascent)
    Form Liquid
    Product Line Alexa Fluor, Click-iT
    Quantity 1 kit
    Emission Wavelength Visible
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.