missing translation for 'onlineSavingsMsg'
Learn More

Clevudine, Thermo Scientific™

Catalog No. p-7238603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469122500 250 mg
AC469120010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469122500 Supplier Thermo Scientific Chemicals Supplier No. 469122500
Only null left

Chemical Identifiers

CAS 163252-36-6
Molecular Formula C10H13FN2O5
Molecular Weight (g/mol) 260.22
MDL Number MFCD00935785
InChI Key GBBJCSTXCAQSSJ-XQXXSGGOSA-N
IUPAC Name 1-[(2S,3R,4S,5S)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN([C@H]2O[C@@H](CO)[C@H](O)[C@H]2F)C(=O)NC1=O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Light Yellow
Quantity 250 mg
Chemical Name or Material Clevudine
Hazard Category Exclamation mark
Recommended Storage Normal conditions