missing translation for 'onlineSavingsMsg'
Learn More

Clenbuterol hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. C0801ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0801ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0801ML Supplier MilliporeSigma Supplier No. C0801ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Clenbuterol is a pharmaceutical decongestant and bronchodilator prescribed to treat asthma with illicit uses as a performance-enhancing drug.
  • This Snap-N-Spike™ Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and urine drug testing and to clinical toxicology and sports testing.

Specifications

Chemical Name or Material Clenbuterol HCl
Packaging Ampule
CAS 21898-19-1
Molecular Formula C12H19Cl3N2O
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Pharmaceutical
Molecular Weight (g/mol) 313.65
Quantity 1 mL
Grade Certified
Show More Show Less