missing translation for 'onlineSavingsMsg'
Learn More

Clenbuterol-d9 hydrochloride, Analytical Standard, ≥97.0% (isotopic purity), MilliporeSigma™ Supelco™

Catalog No. 111017955
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017955 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017955 Supplier Merck Emd Millipore Supplier No. 5496910MG

Clenbuterol-d9 hydrochloride may be used as an internal standard for the quantification of clenbuterol in biological samples by ultra-high pressure liquid chromatography-tandem mass spectrometry (UPLC-MS/MS).

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 184006-60-8
Physical Form Neat
Linear Formula C12D9H9Cl2N2O · HCl
MDL Number MFCD16294267
Synonym 1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butyl-d9-amino)ethanol hydrochloride
Quantity 10 mg
Formula Weight 322.71
Percent Purity ≥97.0% (isotopic purity)
Grade Analytical Standard