missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Clemastine fumarate
$461.88 - $1315.20
Chemical Identifiers
| CAS | 14976-57-9 |
|---|---|
| Molecular Formula | C25H30ClNO5 |
| Molecular Weight (g/mol) | 459.97 |
| InChI Key | PMGQWSIVQFOFOQ-YKVZVUFRSA-N |
| IUPAC Name | (2E)-but-2-enedioic acid; (2R)-2-{2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl}-1-methylpyrrolidine |
| SMILES | OC(=O)\C=C\C(O)=O.CN1CCC[C@@H]1CCO[C@](C)(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465422500
|
Thermo Scientific Chemicals
465422500 |
250 mg |
Each for $461.88
|
|
|||||
|
AC465420010
|
Thermo Scientific Chemicals
465420010 |
1 g |
Each for $1,315.20
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14976-57-9 | |
| 459.97 | |
| (2E)-but-2-enedioic acid; (2R)-2-{2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl}-1-methylpyrrolidine |
| C25H30ClNO5 | |
| PMGQWSIVQFOFOQ-YKVZVUFRSA-N | |
| OC(=O)\C=C\C(O)=O.CN1CCC[C@@H]1CCO[C@](C)(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 |
Specifications
| 14976-57-9 | |
| C25H30ClNO5 | |
| PMGQWSIVQFOFOQ-YKVZVUFRSA-N | |
| (2E)-but-2-enedioic acid; (2R)-2-{2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl}-1-methylpyrrolidine | |
| 459.97 |
| 158.0°C to 162.0°C | |
| 250 mg | |
| OC(=O)\C=C\C(O)=O.CN1CCC[C@@H]1CCO[C@](C)(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 | |
| 459.97 | |
| Clemastine fumarate |
Safety and Handling
EINECSNumber : 239-055-2
RUO – Research Use Only