accessibility menu, dialog, popup

UserName

CH3N(R)O(R)CH3ClOHOOHO(E)

CAS RN 14976-57-9

IUPAC Name: (2E)-but-2-enedioic acid; (2R)-2-{2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl}-1-methylpyrrolidine
Synonyms: clemastine hydrogen fumarate
Molecular Weight (g/mol): 459.97
Molecular Formula: C25H30ClNO5
InChi Key: PMGQWSIVQFOFOQ-YKVZVUFRSA-N
SMILES: OC(=O)\C=C\C(O)=O.CN1CCC[C@@H]1CCO[C@](C)(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results