missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Clemastine fumarate
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 158.0°C to 162.0°C |
| Quantity | 250 mg |
| Formula Weight | 459.97 |
| Chemical Name or Material | Clemastine fumarate |
Safety and Handling
| EINECSNumber | 239-055-2 |
RUO – Research Use Only