missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLEC4F Monoclonal Antibody (370901)

Catalog No. PIMA524113
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524113 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524113 Supplier Invitrogen™ Supplier No. MA524113
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CLECSF4F, rhCLEC4D, rhCLEC4E, recombinant mouse (rm) OCIL, and rmOCIL-rp2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Receptor with an affinity for galactose and fucose. Could be involved in endocytosis. Target protein's carbohydrate binding receptor with an affinity for fucose and galactose.

Specifications

Antigen CLEC4F
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 370901
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Clec4f
Gene Accession No. P70194
Gene Alias Clec4f; Clecsf13; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 13; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13; C-type lectin 13; C-type lectin domain family 4 member F; C-type lectin domain family 4, member f; C-type lectin superfamily member 13; D18063; KCLR; KCR; KUCR_MOUSE; Kupffer cell c-type lectin receptor; kupffer cell receptor
Gene Symbols Clec4f
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CLEC4F/CLECSF13 Ala65-Gly548.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51811
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less