missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Clear Serum Bottle, Crimp Top
Constructed of borosilicate glass, closures are not included
$103.38 - $701.00
Specifications
| Description | No Closure Included |
|---|---|
| Color | Clear |
| Material | Glass |
| Closure Type | Crimp Cap |
| Autoclavable | Autoclavable |
| Catalog Number | Mfr. No. | Volume (Metric) | No. per Pack | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | No. per Pack | Price | Quantity & Availability | ||||
|
111011544
|
MilliporeSigma™ Supelco™
33106 |
30 mL | 36/Pk. |
Each for $103.38
|
|
||||
|
111016329
|
MilliporeSigma™ Supelco™
33109U |
50 mL | 272/Pk. |
Each for $603.00
|
|
||||
|
111010959
|
MilliporeSigma™ Supelco™
33110U |
100 mL | 36/Pk. |
Each for $154.54
|
|
||||
|
111009449
|
MilliporeSigma™ Supelco™
33111U |
100 mL | 144/Pk. |
Each for $504.00
|
|
||||
|
111009575
|
MilliporeSigma™ Supelco™
33107 |
30 mL | 288/Pk. |
Each for $701.00
|
|
||||
Specifications
| No Closure Included | |
| Glass | |
| Autoclavable |
| Clear | |
| Crimp Cap | |
| Serum Bottle |