Learn More
Fisherbrand™ Clear Boston Round Glass Bottles without Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage
 View more versions of this product
Catalog No. FB02911944
Only null left
Description
Description
  • General use bottles ideal for when content integrity and sample or product visibility is important
  • Narrow screw neck opening, curved shoulders, and a round body
  • Clear glass offers minimal chemical interference with contents and is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Specifications
Chemical Identifiers
Clear
No
Boston Round Bottle
Glass (Type 3)
Narrow
Specifications
60 mL
Clear
10 mm
288
No
94 mm
20-400 mm
2 oz.
18 mm
Glass (Type 3)
20-400
Narrow
Boston Round Bottle
39 mm
Product Suggestions
Product Suggestions