Learn More
Fisherbrand™ Cleanroom Pens
Catalog No. 18001507
Change view
Click to view available options
Color:
Black
Catalog No. Color
18001507 Black
1 options
Catalog No. 18001507 Supplier Fisherbrand™ Supplier No. 18001507
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed with low sodium ink to prevent contamination in cleanroom settings. Fisherbrand™ Cleanroom Pens come in black, blue, and red and feature HDPE construction.
  • Manufactured with very low sodium inks to prevent contamination
  • Barrel and cap made from high density polyethylene (HDPE)
  • Pens are packaged 10 per pack
  • Available in Black (18-001-507), Blue (18-001-511), or Red (18-001-508)
Specifications
Color Black