Learn More
Fisherbrand™ Cleanroom Notebooks, Latex
Catalog No. 18001544
Change view
Click to view available options
Dimensions (L x W):
5.5 x 8.5 in.
5.875 x 8.25 in
8.25 x 11.75 in.
3.5 x 5 in.
8.5 x 11 in.
3 x 5 in.
Ruled:
College Ruled
Grid
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Ruled
18001544 8.5 x 11 in. Grid
18003063 8.25 x 11.75 in. College Ruled
18003064 5.875 x 8.25 in College Ruled
18001500 8.5 x 11 in. College Ruled
18001547 3.5 x 5 in. Grid
18001501 5.5 x 8.5 in. College Ruled
18001502 3 x 5 in. College Ruled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18001544 Supplier Fisherbrand™ Supplier No. 01NB8.5X11GRID
Only null left
European sizing cleanroom notebooks, college ruled.
  • For documentation in critical environments
  • Low particulate and ESD generation
  • Printed on #22 paper with college rule lines.
  • Semi-transparent high density polyethylene (HDPE) covers
  • 100 pages/50 sheets
  • Spiral bound
Compliance
Class 10 Certification
Specifications
Material Latex
Ruled Grid
Dimensions (L x W) 8.5 x 11 in.
Product Type Notebook
Weight (English) 22#