missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cleanroom Click Pen

Catalog No. 18001510
Change view
Click to view available options
Color:
Black Ink
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18001510 Black Ink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18001510 Supplier Fisherbrand™ Supplier No. 18001510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prevent the possibility of dropping or loosing the pen cap. Fisherbrand Cleanroom Click Pens are an alternative to cap pens.

  • Nonsterile
  • Class 10 certified
  • Manufactured with very low sodium to prevent sodium contamination
  • Barrel and clicker are made from high-density polyethylene
  • Suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms

Specifications

Material HDPE Barrel and Clicker
Color Black Ink
Description No cap
Certifications/Compliance Class 10
Product Type Cleanroom Click Pen