Learn More
New Pig CleanMax™ Cleanroom Sterile Coverall (Elastic Wrists and Ankles)
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile coverall with elastic wrists and ankles
$1150.34 - $1354.85
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-987-925
View Documents
New Pig
WPL30064XL
4X-Large
Case of 25 Each for $1,303.72
Only null left
17-987-926
View Documents
New Pig
WPL30065XL
5X-Large
Case of 25 Each for $1,354.85
Only null left
17-987-921
View Documents
New Pig
WPL3006L
Large
Case of 25 Each for $1,150.34
Only null left
17-987-920
View Documents
New Pig
WPL3006M
Medium
Case of 25 Each for $1,150.34
Only null left
17-987-919
View Documents
New Pig
WPL3006S
Small
Case of 25 Each for $1,150.34
Only null left
17-987-922
View Documents
New Pig
WPL3006XL
X-Large
Case of 25 Each for $1,150.34
Only null left
17-987-923
View Documents
New Pig
WPL3006XXL
2X-Large
Case of 25 Each for $1,201.47
Only null left
17-987-924
View Documents
New Pig
WPL3006XXXL
3X-Large
Case of 25 Each for $1,252.60
Only null left
Description
Description
  • A key feature is its sterile status, as it has been gamma-irradiated to a Sterility Assurance Level (SAL) of -6.
  • The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • Despite its strong barrier properties, the fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
  • The sterile coveralls are typically folded in a specific manner to facilitate aseptic donning, minimizing the risk of contamination during the gowning process. They are also double-bagged and packaged in a cleanroom environment.
  • The coverall includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles with 25 per cases to provide a secure fit and prevent the ingress of contaminants.