Learn More
New Pig CleanMax™ Cleanroom Sterile Coverall (Elastic Wrists and Ankles)
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile coverall with elastic wrists and ankles
Supplier:  New Pig WPL3006L
 View more versions of this product
Catalog No. 17-987-921
Only null left
Explore available promotions
Description
Description
  • A key feature is its sterile status, as it has been gamma-irradiated to a Sterility Assurance Level (SAL) of -6.
  • The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • Despite its strong barrier properties, the fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
  • The sterile coveralls are typically folded in a specific manner to facilitate aseptic donning, minimizing the risk of contamination during the gowning process. They are also double-bagged and packaged in a cleanroom environment.
  • The coverall includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles with 25 per cases to provide a secure fit and prevent the ingress of contaminants.
Specifications
Specifications
Large
Product Suggestions
Product Suggestions