Learn More
New Pig CleanMax™ Cleanroom Coverall (Elastic wrists and Ankles)
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this coverall with elastic wrists and ankles
Supplier:  New Pig WPL3005M
 View more versions of this product
Catalog No. 17-987-912
Only null left
Explore available promotions
Description
Description
  • Constructed from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven.
  • The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • Provides enhanced strength and abrasion resistance, ensuring the coverall can withstand the rigors of use without compromising its protective qualities.
  • Fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
  • The coverall includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles to provide a secure fit and prevent the ingress of contaminants.
  • The CleanMax™ product line is often positioned as an economical alternative to more expensive cleanroom garments, providing a high level of protection at a lower cost for applications that require a balance of performance and value.
Specifications
Specifications
Medium
Product Suggestions
Product Suggestions