Learn More
New Pig CleanMax™ Cleanroom Coverall (Elastic wrists and Ankles)
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this coverall with elastic wrists and ankles
$1035.66 - $1242.79
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-987-917
View Documents
New Pig
WPL30054XL
4X-Large
Case of 25 Each for $1,191.00
Only null left
17-987-918
View Documents
New Pig
WPL30055XL
5X-Large
Case of 25 Each for $1,242.79
Only null left
17-987-913
View Documents
New Pig
WPL3005L
Large
Case of 25 Each for $1,035.66
Only null left
17-987-912
View Documents
New Pig
WPL3005M
Medium
Case of 25 Each for $1,035.66
Only null left
17-987-911
View Documents
New Pig
WPL3005S
Small
Case of 25 Each for $1,035.66
Only null left
17-987-914
View Documents
New Pig
WPL3005XL
X-Large
Case of 25 Each for $1,035.66
Only null left
17-987-915
View Documents
New Pig
WPL3005XXL
2X-Large
Case of 25 Each for $1,087.43
Only null left
17-987-916
View Documents
New Pig
WPL3005XXXL
3X-Large
Case of 25 Each for $1,139.22
Only null left
Description
Description
  • Constructed from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven.
  • The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • Provides enhanced strength and abrasion resistance, ensuring the coverall can withstand the rigors of use without compromising its protective qualities.
  • Fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
  • The coverall includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles to provide a secure fit and prevent the ingress of contaminants.
  • The CleanMax™ product line is often positioned as an economical alternative to more expensive cleanroom garments, providing a high level of protection at a lower cost for applications that require a balance of performance and value.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'