Learn More
New Pig CleanMax™ Cleanroom Coverall (Bound Seams and Attached Hood)
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this coverall with zipper front and elastic hood
$945.83 - $1150.34
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-987-933
View Documents
New Pig
WPL30074XL
4X-Large
Case of 25 Each for $1,099.22
Only null left
17-987-934
View Documents
New Pig
WPL30075XL
5X-Large
Case of 25 Each for $1,150.34
Only null left
17-987-929
View Documents
New Pig
WPL3007L
Large
Case of 25 Each for $945.83
Only null left
17-987-928
View Documents
New Pig
WPL3007M
Medium
Case of 25 Each for $945.83
Only null left
17-987-927
View Documents
New Pig
WPL3007S
Small
Case of 25 Each for $945.83
Only null left
17-987-930
View Documents
New Pig
WPL3007XL
X-Large
Case of 25 Each for $945.83
Only null left
17-987-931
View Documents
New Pig
WPL3007XXL
2X-Large
Case of 25 Each for $996.96
Only null left
17-987-932
View Documents
New Pig
WPL3007XXXL
3X-Large
Case of 25 Each for $1,048.09
Only null left
Description
Description
  • Bound seams and a laminated fabric provide a strong barrier to both particles and non-hazardous liquids.
  • The attached hood offers complete head-to-toe coverage, which is essential for maintaining a cleanroom environment.
  • It's low-linting, which helps prevent contamination by shedding very few particles or fibers.
  • A zippered front closure with a storm flap and elastic wrists and ankles ensure a secure fit, sealing out contaminants.
  • The fabric is breathable, allowing moisture vapor to escape and reducing heat stress on the wearer.
  • This coverall is a cost-effective choice that balances performance with value, making it suitable for a range of controlled environments.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'