Learn More
New Pig CleanMax™ Cleanroom Coverall (Bound Seams and Attached Hood)
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this coverall with zipper front and elastic hood
Supplier:  New Pig WPL30074XL
 View more versions of this product
Catalog No. 17-987-933
Only null left
Explore available promotions
Description
Description
  • Bound seams and a laminated fabric provide a strong barrier to both particles and non-hazardous liquids.
  • The attached hood offers complete head-to-toe coverage, which is essential for maintaining a cleanroom environment.
  • It's low-linting, which helps prevent contamination by shedding very few particles or fibers.
  • A zippered front closure with a storm flap and elastic wrists and ankles ensure a secure fit, sealing out contaminants.
  • The fabric is breathable, allowing moisture vapor to escape and reducing heat stress on the wearer.
  • This coverall is a cost-effective choice that balances performance with value, making it suitable for a range of controlled environments.
Specifications
Specifications
4X-Large
Product Suggestions
Product Suggestions