missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Cleaned Copper Tubing, Length: 50 ft.
Solvent-washed to ASTM B-280 specifications
$244.76 - $391.06
Specifications
| Description | Solvent-Washed to ASTM B-280 Specifications |
|---|---|
| For Use With (Application) | GC Plumbing Applications |
| Overall Length (English) | 50 ft. |
| Material | Copper |
Specifications
| Solvent-Washed to ASTM B-280 Specifications | |
| GC Plumbing Applications | |
| 50 ft. | |
| Copper |