missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 28# Latex Free Cleanroom Paper

Catalog No. 18001549
Change view
Click to view available options
Color:
Blue
Canary
Green
Pink
Dimensions (L x W):
8.5 x 11 in.
11 x 17 in.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
18001549 Blue 11 x 17 in.
18001562 Blue 8.5 x 11 in.
18001553 Canary 11 x 17 in.
18001551 Canary 8.5 x 11 in.
18001555 Green 11 x 17 in.
18001559 Green 8.5 x 11 in.
18001557 Pink 11 x 17 in.
18001561 Pink 8.5 x 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18001549 Supplier Fisherbrand™ Supplier No. PA28LFBLU17

Designed for writing, printing and photocopying in cleanroom environments. Fisherbrand cleanroom paper is ideal for documentation in critical environments, where very low particle and ESD generation is important. This clean room product is double bagged and 'Class 10 Certified.'

  • Clean Room
  • Latex-Free

Compliance

Class 10 Certification

Specifications

Material Latex-free
Color Blue
Dimensions (L x W) 11 x 17 in.
Plain paper Yes
Weight (English) 28#