missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLCNKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103736 Supplier Invitrogen™ Supplier No. PA5103736
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLCNKB.

Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney.

Specifications

Antigen CLCNKB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLCNKB
Gene Accession No. P51801
Gene Alias chloride channel; chloride channel K1-like; putative basolateral cTAL chloride channel ClC-Ka; chloride channel K2; chloride channel Ka; Chloride channel Kb; chloride channel protein ClC-Ka; Chloride channel protein ClC-Kb; chloride channel, kidney, B; chloride channel, voltage-sensitive Ka; chloride channel, voltage-sensitive Kb; chloride voltage-gated channel Kb; clC-K1; Clck2; ClC-K2; ClC-K2L; Clc-Ka; CLCKB; ClC-Kb; CLCNK; Clcnk1l; CLCNKA; Clcnkb; hClC-Kb; LOC100730738; MGC24087; putative basolateral mTAL chloride channel ClC-Ka; RP11-5P18.8
Gene Symbols CLCNKB
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLCNKB(Accession P51801), corresponding to amino acid residues G6-L56.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1188
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less