missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-Ethylcarbodiimide Hydrochloride, Spectrum™ Chemical

Catalog No. 18620622
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18620622 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18620622 Supplier Spectrum Chemical Mfg Cor Supplier No. D2190500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Synonym: EDAC, 1-Ethyl-3-(3-Dimethylaminopropyl)Carbodiimide Hydrochloride

Chemical Identifiers

CAS 25952-53-8

Specifications

Quantity 500 g
Molecular Weight (g/mol) 191.71
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride