missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Pellets, BiotechGrade, 95-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Hydroxide, Pellets, BiotechGrade or caustic soda, is an inorganic, highly basic compound that is water soluble, as well as soluble in ethanol and methanol. One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Specifications
Specifications
| Color | White |
| CAS Max % | 1 |
| Quantity | 50 kg |
| Percent Purity | 95 to 100.5% |
| Grade | Biotech |
| Odor | Odorless |
| Packaging | Fiber Drum |
| Physical Form | Pellet |
| Chemical Name or Material | Sodium Hydroxide |