Learn More
Eisco™ Class B Volumetric Pipets
The bulb form class B pipette is made of borosilicate glass and is ideal for pipetting liquid substances from one vessel to another with accuracy. Recommended for use with a pipet pump.
Supplier:  Eisco™ CH0558A
 View more versions of this product
Catalog No. S89319
Only null left
Explore available promotions
Description
Description
  • Made from 3.3 borosilicate glass
  • Blue graduations
  • Bulb form
Specifications
Specifications
No
DIN ISO 648
Class B
Non-sterile
Yes
Blue
1
0.03 oz.
1 mL
Serological Pipet
Autoclavable
B
Borosilicate Glass
Pipetting liquid substances from one vessel to another with accuracy
Blue
12 in.
±0.015 mL
Blue
30.48 cm
Reusable
Product Suggestions
Product Suggestions