Learn More
Eisco™ Class B Round Base Graduated Cylinder
Eisco™ Class B Round Base Graduated Cylinder feature printed graduations and spouts.
$15.04 - $226.43
Specifications
Base Type Round
Class Class B
Type Graduated Measuring Cylinder
Scale Color Blue
Shape Round
View More Specs
Products 10
Catalog Number Mfr. No. Capacity (Metric) Graduations Includes Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Includes Price Quantity  
S88426
View Documents
Eisco™
CH0344A5
5 mL 0.1 mL 1 Ea.
Each for $18.68
Only null left
S27993
View Documents
Eisco™
CH0344A10
10 mL 0.2 mL 1 Ea.
Each for $15.04
Only null left
S15486
View Documents
Eisco™
CH0344A10PK12
10 mL 0.2 mL 12/Pk.
Each for $226.43
Only null left
S15455
View Documents
Eisco™
CH0344A
25 mL 0.5 mL 1 Ea.
Each for $20.93
Only null left
S15456
View Documents
Eisco™
CH0344B
50 mL 1 mL 1 Ea.
Each for $28.39
Only null left
S88428
View Documents
Eisco™
CH0344C
100 mL 1 mL 1 Ea.
Each for $27.18
Only null left
S88429
View Documents
Eisco™
CH0344D
250 mL 2 mL 1 Ea.
Each for $31.57
Only null left
S88430
View Documents
Eisco™
CH0344E
500 mL 5 mL 1 Ea.
Each for $57.86
Only null left
S88431
View Documents
Eisco™
CH0344F
1000 mL 10 mL 1 Ea.
Each for $102.25
Only null left
S88432
View Documents
Eisco™
CH0344G
2000 mL 20 mL 1 Ea.
Each for $163.50
Only null left
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Single piece construction with round base
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class B
  • Conforms to DIN EN ISO 4788 standard
  • Autoclavable
Specifications
Specifications
Round
Graduated Measuring Cylinder
Round
Class B
Blue
Borosilicate Glass