missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Class B Glass serological Pipettes
Color-coded Class B Serological Pipettes are made of high quality Borosilicate 3.3 glass
$14.07 - $18.33
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Calibrated | Yes |
| Calibration | To Deliver |
| Certifications/Compliance | DIN ISO 835 |
| Class | Class B |
Description
- Made from high quality, heavy duty, borosilicate glass
- Accuracy as per Class B
- Color coded as per DIN ISO 835 standards
- Calibrated for delivery to jet
- Last drop expelled by blowing
Specifications
| Autoclavable | |
| To Deliver | |
| Class B | |
| Borosilicate Glass (3.3) | |
| Blue | |
| Serological Pipet |
| Yes | |
| DIN ISO 835 | |
| Class B | |
| Non-sterile | |
| 1 | |
| Reusable |