missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Class B Glass Automatic Schellbach Zero Burettes with Intermediate Stopcock, Reservoir and Bellow

High-quality Class-B automatic zero burets made from borosilicate 3.3 glass. Designed with Schellbach buret, an intermediate PTFE stopcock and rubber bellow, calibrated for delivery. Includes glass reservior.

$646.61 - $743.96

Specifications

Autoclavable Autoclavable
Closure Type Intermediate Stopcock
Color Clear
Closure Material PTFE
For Use With (Equipment) Labware
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S26707
View Documents Promotion Details
Eisco™
CH0242T
10 mL
Each for $646.61
Only null left
 
S26708
View Documents Promotion Details
Eisco™
CH0242U
25 mL
Each for $646.61
Only null left
 
S26710
View Documents Promotion Details
Eisco™
CH0242W
100 mL
Each for $743.96
Only null left
 
Description

Description

  • Automatic zero Schellbach burette
  • Class B
  • Complies with ISO 385
  • Features intermediate PTFE stopcock, reservoir, joint clip, and rubber bellow
  • Made from high quality Borosilicate 3.3 glass
  • Blue enamel graduations with Schellbach stripe (vertical white stripe and central blue ribbon) for accurate reading of meniscus
  • Intermediate stopcock to facilitate drainage or excess liquid back into the reservoir
  • Graduated reservoir
  • Securely packed for safe transit
Specifications

Specifications

Autoclavable
Clear
Labware
Dispensing liquid
PTFE
PTFE
Class B
Intermediate Stopcock
PTFE
Borosilicate Glass (3.3)
Burette, reservoir, bellow, joint clip
Yes
Automatic Self-zeroing Schellbach Buret
DIN ISO 385
Videos
SDS
Documents

Documents