Learn More
Eisco™ Class A Volumetric Pipet, Borosilicate Glass
Eisco Labs Volumetric Pipet, Borosilicate glass, 0.030 tolerance, Class A Glass, Individually wrapped
Supplier:  Eisco™ CH0559F
 View more versions of this product
Catalog No. S89328
Only null left
Explore available promotions
Description
Description
  • Volumetric pipette featuring color coding for easy volume identification.
  • Made of high-quality Borosilicate 3.3 glass.
  • Autoclavable, with superior chemical durability.
  • Low temperature gradient - can withstand high temperatures and thermal shocks.
  • Class A tolerance ±0.010 mL, as per DIN ISO 648 standard.
  • Calibrated for delivery.
  • Each pipette is individually wrapped to keep clean.
Specifications
Specifications
Autoclavable
Non-sterile
0.68 oz.
20 mL
Serological Pipet
Borosilicate Glass
Blue
Yellow
50.8 cm
Reusable
Product Suggestions
Product Suggestions