Learn More
Eisco™ Class A Round Base Glass Graduated Cylinders
Eisco™ Class A Round Base Glass Graduated Cylinders feature printed graduations and spouts.
$13.80 - $148.13
Specifications
| Base Type | Round |
|---|---|
| Certifications/Compliance | DIN EN ISO 4788 |
| Class | A |
| Type | Graduated Measuring Cylinder |
| Shape | Round |
| Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Scale Color | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Scale Color | Price | Quantity | |||
|
S88434
|
Eisco™
CH0344HWT |
5 mL | 0.1 mL | White |
Each for $13.80
|
|
|||
|
S88435
|
Eisco™
CH0344I |
10 mL | 0.2 mL | Blue |
Each for $19.43
|
|
|||
|
S88437
|
Eisco™
CH0344J |
25 mL | 0.5 mL | Blue |
Each for $21.83
|
|
|||
|
S88438
|
Eisco™
CH0344LWT |
100 mL | 1 mL | White |
Each for $16.40
|
|
|||
|
S88439
|
Eisco™
CH0344M |
250 mL | 2 mL | Blue |
Each for $30.10
|
|
|||
|
S88440
|
Eisco™
CH0344N |
500 mL | 5 mL | Blue |
Each for $50.43
|
|
|||
|
S88441
|
Eisco™
CH0344O |
1000 mL | 10 mL | Blue |
Each for $90.57
|
|
|||
|
S88442
|
Eisco™
CH0344OWT |
1000 mL | 10 mL | White |
Each for $81.87
|
|
|||
|
S88443
|
Eisco™
CH0344P |
2000 mL | 10 mL | Blue |
Each for $148.13
|
|
|||
|
S15487
|
Eisco™
CH0344K |
50 mL | 1 mL | Blue |
Each for $27.03
|
|
|||
|
S15488
|
Eisco™
CH0344L |
100 mL | 1 mL | Blue |
Each for $30.30
|
|
|||
Description
- Made from high quality Borosilicate 3.3 Glass
- Single piece construction with round base
- Printed graduations are chemical resistant
- Spout for easy, mess free pouring
- Class A
- Conforms to DIN EN ISO 4788 standard
- Autoclavable
Specifications
| Round | |
| A | |
| Round | |
| Measuring Liquids |
| DIN EN ISO 4788 | |
| Graduated Measuring Cylinder | |
| Borosilicate Glass |