missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Class 2 Laser Pointer

Laser pointer designed with safety in mind.

Supplier:  United Scientific™ RLPCLS2

 View more versions of this product

Catalog No. S00354


Only null left
Explore available promotions

Description

Description

Economical laser pointers are useful for many optical experiments.

  • Ideal for middle school and high school classrooms
  • Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip
  • Class 2 Red laser pointer has a maximum output of 1 mW and a wavelength of 650 nm (± 10 nm)
  • Class 2 Green laser pointer has a maximum output of 1 mW and a wavelength of 532 nm (± 10 nm)
Specifications

Specifications

Laser Pointer
12.7 mm
15.24 cm
1 mW
Beam: 75 m
0.5 in.
6 in.
AAA
Red
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions