missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Class 2 Laser Pointer

Laser pointer designed with safety in mind.

$41.82 - $137.18

Specifications

Product Type Laser Pointer
Diameter (English) 0.5 in.
Diameter (Metric) 12.7 mm
Length (English) 6 in.
Length (Metric) 15.24 cm
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
S00354
View Documents Promotion Details
United Scientific™
RLPCLS2
Red
Each for $41.82
Only null left
 
S00355
View Documents Promotion Details
United Scientific™
GLPCLS2
Green
Each for $137.18
Only null left
 
Description

Description

Economical laser pointers are useful for many optical experiments.

  • Ideal for middle school and high school classrooms
  • Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip
  • Class 2 Red laser pointer has a maximum output of 1 mW and a wavelength of 650 nm (± 10 nm)
  • Class 2 Green laser pointer has a maximum output of 1 mW and a wavelength of 532 nm (± 10 nm)
Specifications

Specifications

Laser Pointer
12.7 mm
15.24 cm
1 mW
0.5 in.
6 in.
AAA
Videos
SDS
Documents

Documents