Learn More
National Safety Apparel™ ArcGuard™ Class 00 Rubber Voltage Gloves Premium Kit
Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 00 insulating gloves will protect workers from up to 500 V.
Supplier: National Safety Apparel™ KITGC0010AG
Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- FR knit gloves stretch for dexterity and can be worn for another layer of protection
- Compliant with ANSI/ASTM D120 and NFPA 70E standards
- Class 00 gloves offer protection from up to 500 V
- Gloves should be used within one year of marked test date
- When not in use, lay flat in the included protective bag
- Store away from direct sunlight, high heat, and humidity
Specifications
| ANSI/ASTM D120, Class 00, Type I, NFPA 70E, ASTM F1506, OSHA 1910.269 | |
| Black | |
| Electrical Insulating Gloves Kit | |
| Electrical Utility Work, Shock Protection | |
| Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag | |
| 500 V |
| Industrial | |
| Rubber, Leather | |
| 10 | |
| Rolled | |
| KITGC00BAG |