missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ ArcGuard™ Class 00 Rubber Voltage Gloves Premium Kit
Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- FR knit gloves stretch for dexterity and can be worn for another layer of protection
- Compliant with ANSI/ASTM D120 and NFPA 70E standards
- Class 00 gloves offer protection from up to 500 V
- Gloves should be used within one year of marked test date
- When not in use, lay flat in the included protective bag
- Store away from direct sunlight, high heat, and humidity
Specifications
Specifications
| Certifications/Compliance | ANSI/ASTM D120, Class 00, Type I, NFPA 70E, ASTM F1506, OSHA 1910.269 |
| Applications | Industrial |
| Color | Black |
| Material | Rubber, Leather |
| Product Type | Electrical Insulating Gloves Kit |
| Size | 8 |
| For Use With (Application) | Electrical Utility Work, Shock Protection |
| Glove Cuff Style | Rolled |
| Includes | Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag |
| Series | KITGC00BAG |
| Show More |