missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLASP1 Recombinant Rabbit Monoclonal Antibody (23GB3810)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | CLASP1 |
|---|---|
| Clone | 23GB3810 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
Description
CLASP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.Specifications
| CLASP1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CLASP1 | |
| 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1 | |
| CLASP1 | |
| Primary | |
| Affinity chromatography |
| 23GB3810 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7Z460, Q80TV8 | |
| 23332, 304740, 76707 | |
| A synthetic peptide of human CLASP1 (1-52AA). | |
| Antibody |