missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLASP1 Recombinant Rabbit Monoclonal Antibody (23GB3810)

Description
CLASP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
Specifications
Specifications
| Antigen | CLASP1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB3810 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CLASP1 |
| Gene Accession No. | Q7Z460, Q80TV8 |
| Gene Alias | 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1 |
| Gene Symbols | CLASP1 |
| Show More |