missing translation for 'onlineSavingsMsg'
Learn More

Clarithromycin 98.0+%, TCI America™

Catalog No. C22205G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22205G 5 g
C22201G 1 g
C222025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C22205G Supplier TCI America Supplier No. C22205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81103-11-9
Molecular Formula C38H69NO13
Molecular Weight (g/mol) 747.964
MDL Number MFCD00865140
InChI Key AGOYDEPGAOXOCK-KCBOHYOISA-N
Synonym clarithromycin, biaxin, clarithromycine, 6-o-methylerythromycin, clathromycin, klaricid, 6-o-methylerythromycin a, abbott-56268, macladin, clarith
PubChem CID 84029
ChEBI CHEBI:3732
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion
SMILES CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O

Specifications

Melting Point 225°C
Color White
Quantity 5 g
Formula Weight 747.96
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Clarithromycin
RTECSNumber KF4997000
TSCA No