missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clarithromycin 98.0+%, TCI America™
Supplier: TCI America C222025G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 81103-11-9 | |
| 747.964 | |
| AGOYDEPGAOXOCK-KCBOHYOISA-N | |
| 84029 | |
| (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion |
| C38H69NO13 | |
| MFCD00865140 | |
| clarithromycin, biaxin, clarithromycine, 6-o-methylerythromycin, clathromycin, klaricid, 6-o-methylerythromycin a, abbott-56268, macladin, clarith | |
| CHEBI:3732 | |
| CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O |
Specifications
| Clarithromycin | |
| 225°C | |
| C38H69NO13 | |
| MFCD00865140 | |
| AGOYDEPGAOXOCK-KCBOHYOISA-N | |
| (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion | |
| 84029 | |
| 747.96 | |
| Crystalline Powder |
| 81103-11-9 | |
| White | |
| 25 g | |
| clarithromycin, biaxin, clarithromycine, 6-o-methylerythromycin, clathromycin, klaricid, 6-o-methylerythromycin a, abbott-56268, macladin, clarith | |
| CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O | |
| 747.964 | |
| CHEBI:3732 | |
| ≥98.0% (T) |
Safety and Handling
RTECSNumber : KF4997000
TSCA : No