missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clarithromycin Impurity Q Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 118074-07-0 |
| Linear Formula | C38H69 NO14 |
| MDL Number | MFCD24386630 |
| Synonym | Clarithromycin N-oxide; 6-O-Methylerythromycin N-oxide |
| Type | Certified Reference Material |
| Quantity | 20 mg |
| Formula Weight | 763.95 |
| Grade | Certified Reference Material |