Learn More
Clarithromycin, 960-1040 ^mg/mg
CAS: 81103-11-9 | C38H69NO13 | 747.964 g/mol
Supplier: Thermo Scientific Chemicals J6600506
| Quantity | 5 g |
|---|
Description
A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
Chemical Identifiers
| 81103-11-9 | |
| 747.964 | |
| AGOYDEPGAOXOCK-KCBOHYOISA-N | |
| CHEBI:3732 | |
| CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O |
| C38H69NO13 | |
| MFCD00865140 | |
| 84029 | |
| (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion |
Specifications
| Clarithromycin | |
| Powder | |
| 81103-11-9 | |
| White | |
| >110°C (230°F) | |
| MFCD00865140 | |
| 14,2339 | |
| AGOYDEPGAOXOCK-KCBOHYOISA-N | |
| (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion | |
| 84029 | |
| 747.95 |
| Clarithromycin | |
| 960 to 1040μg/mg | |
| 217°C to 220°C | |
| 5 g | |
| C38H69NO13 | |
| 86°C | |
| Soluble in DMSO | |
| CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O | |
| 747.964 | |
| CHEBI:3732 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : KF4997000
TSCA : No
Recommended Storage : Ambient temperatures